Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09753

Sigma-Aldrich

Albumin fluorescence assay kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352103

storage temp.

2-8°C

General description

The components can also be ordered separately under the product numbers indicated.

Components

Composition:
05497 Albumin Blue 580 Potassium salt Solution in 2-propanol, 10 mL
79438 Buffer for the determination of Albumine, 2×250 mL
05418 Albumin from human serum (HSA), 2×1 g
05476 Albumin from bovine serum (BSA), 10 g
09761 Calibrator Diluent Solution, 100 ml

Other Notes

For the specific and sensitive spectrofluorometric determination of albumine (1-200 mg/l) in natural matrices, using Albumin Blue 580[1]

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Albumin blue 580 fluorescence assay for albumin.
M A Kessler et al.
Analytical biochemistry, 248(1), 180-182 (1997-05-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service