Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09729

Sigma-Aldrich

Ammonium iron(III) sulfate dodecahydrate

BioUltra, ≥99.0% (RT)

Synonym(s):

Ammonium ferric sulfate dodecahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Fe(SO4)2 · 12H2O
CAS Number:
Molecular Weight:
482.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioUltra

assay

≥99.0% (RT)

impurities

insoluble matter, passes filter test

pH

~1.0 (25 °C, 1 M in H2O)

mp

39-41 °C (lit.)

solubility

H2O: 1 m at 20 °C, clear, yellow-brown to red-brown

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤100 mg/kg
phosphate (PO43-): ≤30 mg/kg

cation traces

As: ≤0.05 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤10 mg/kg
Cr: ≤50 mg/kg
Cu: ≤5 mg/kg
Fe2+: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤10 mg/kg
Mn: ≤50 mg/kg
Na: ≤50 mg/kg
Ni: ≤10 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

N.O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].OS([O-])(=O)=O.[O-]S([O-])(=O)=O

InChI

1S/Fe.H3N.2H2O4S.12H2O/c;;2*1-5(2,3)4;;;;;;;;;;;;/h;1H3;2*(H2,1,2,3,4);12*1H2/q+3;;;;;;;;;;;;;;;/p-3

InChI key

LCPUDZUWZDSKMX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium ferric sulfate dodecahydrate has been used to develop an effective method for preparing magnetic multi-walled carbon nanotubes (M-MWCNTs).

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Wittmann et al.
Chemico-biological interactions, 138(2), 171-187 (2001-10-24)
Diabetes mellitus is characterized by increased methylglyoxal (MG) production. The aim of the present study was to investigate the role of iron in the cellular and molecular effects of MG. A red blood cell (RBC) model and L-arginine were used
Jun-Beum Lee et al.
Bioscience, biotechnology, and biochemistry, 68(3), 495-500 (2004-04-02)
Efficiency was examined of microencapsulating L-ascorbic acid by polyglycerol monostearate (PGMS), and changes in the chemical and sensorial aspects of L-ascorbic acid and/or iron-fortified milk during storage were evaluated. The selected core materials were ferric ammonium sulfate and L-ascorbic acid.
M X Yang et al.
Archives of biochemistry and biophysics, 324(2), 282-292 (1995-12-20)
The NADH-dependent microsomal electron transfer system consists of NADH-cytochrome b5 reductase and cytochrome b5, which donates reducing equivalents to fatty acyl desaturase, cytochrome P450, and other reactions. A study was carried out to investigate the interaction of NADH with several
H S Kwak et al.
Journal of agricultural and food chemistry, 51(26), 7770-7774 (2003-12-11)
This study was designed to develop a microencapsulated iron that could be used to fortify milk and to determine the sensory properties of milk fortified with microencapsulated iron. Coating material was polyglycerol monostearate (PGMS), and selected core material was ferric
Luigi Brunetti et al.
Neurobiology of aging, 25(1), 125-129 (2003-12-17)
In order to investigate whether amyloid beta-peptide-induced oxidative damage in the brain could be related to aging, we studied the release of 8-iso-prostaglandin (PG)F2alpha, a stable marker of cellular oxidative stress, in brain synaptosomes from Wistar rats of different ages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service