Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09715

Sigma-Aldrich

Ammonium bromide

puriss. p.a., ACS reagent, ≥99.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Br
CAS Number:
Molecular Weight:
97.94
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38050407
PubChem Substance ID:
NACRES:
NB.24

grade

ACS reagent
puriss. p.a.

Quality Level

vapor pressure

1 mmHg ( 198.3 °C)

assay

≥99.0% (AT)

form

powder or crystals

pH

5.0-6.0 (25 °C, 50 mg/mL in H2O)

mp

452 °C (lit.)

density

2.43 g/mL at 25 °C (lit.)

anion traces

bromate (BrO3-): ≤10 mg/kg
chloride (Cl-): ≤1000 mg/kg
iodide (I-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg
heavy metals: ≤5 ppm (by ICP-OES)

SMILES string

N.Br[H]

InChI

1S/BrH.H3N/h1H;1H3

InChI key

SWLVFNYSXGMGBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Lact. - Repr. 1B - STOT RE 1 - STOT SE 3

target_organs

Central nervous system, Nervous system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Vibrational Spectra of Molecules and Complex Ions in Crystals IV. Ammonium Bromide and Deutero-Ammonium Bromide.
Wagner WL and Hornig DF.
J. Chem. Phys. , 18(3), 305-312 (1950)
Crystal Structure of the Low Temperature Modification ol Ammonium Bromide.
Ketelaar JAA.
Nature, 134, 250-251 (1934)
Özge Karayel et al.
Scientific reports, 8(1), 2269-2269 (2018-02-06)
The successful completion of cytokinesis requires the coordinated activities of diverse cellular components including membranes, cytoskeletal elements and chromosomes that together form partly redundant pathways, depending on the cell type. The biochemical analysis of this process is challenging due to its
Neutron Diffraction Determination of the Crystal Structure of Ammonium Bromide in Four Phases1.
Levy HA and Peterson SW.
Journal of the American Chemical Society, 75(7), 1536-1542 (1953)
The crystal structure of the ammonium bromide complex of 1, 4, 7, 10, 13, 16-hexaoxacyclooctadecane (18-crown-6), C12H24O6 NH4Br 2H2O.
Nagano O, et al.
Bulletin of the Chemical Society of Japan, 51(3), 790-793 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service