Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09546

Sigma-Aldrich

4-Fluorophenyl phenyl ether

≥98.0%

Synonym(s):

4-Fluorodiphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9FO
CAS Number:
Molecular Weight:
188.20
Beilstein/REAXYS Number:
2047183
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.0%

refractive index

n20/D 1.557

bp

147-149 °C/53 mmHg (lit.)

density

1.155 g/mL at 20 °C (lit.)

SMILES string

Fc1ccc(Oc2ccccc2)cc1

InChI

1S/C12H9FO/c13-10-6-8-12(9-7-10)14-11-4-2-1-3-5-11/h1-9H

InChI key

AODSTUBSNYVSSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

253.4 °F - closed cup

flash_point_c

123 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carsten Ginsel et al.
Drug metabolism and disposition: the biological fate of chemicals, 46(10), 1396-1402 (2018-07-27)
The mitochondrial amidoxime reducing component is a recently discovered molybdenum enzyme in mammals which, in concert with the electron transport proteins cytochrome b5 and NADH cytochrome b5 reductase, catalyzes the reduction of N-oxygenated structures. This three component enzyme system plays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service