Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

09440

Sigma-Aldrich

(R)-(+)-Tetrahydro-3-furylamine p-toluenesulfonate salt

≥99.0% (T)

Synonym(s):

(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO · C7H8O3S
CAS Number:
Molecular Weight:
259.32
Beilstein/REAXYS Number:
5783386
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥99.0% (T)

optical activity

[α]20/D +4.5±0.5°, c = 3% in methanol

mp

139-141 °C

functional group

ether
tosylate

SMILES string

N[C@@H]1CCOC1.Cc2ccc(cc2)S(O)(=O)=O

InChI

1S/C7H8O3S.C4H9NO/c1-6-2-4-7(5-3-6)11(8,9)10;5-4-1-2-6-3-4/h2-5H,1H3,(H,8,9,10);4H,1-3,5H2/t;4-/m.1/s1

InChI key

BZXPLADBSZWDIH-FZSMXKCYSA-N

Other Notes

Chiral amine; amidation with amino acids[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Barlos et al.
Liebigs Ann. Chem., 1950-1950 (1986)
Glorymar Ibanez et al.
Combinatorial chemistry & high throughput screening, 15(5), 359-371 (2012-01-20)
Protein methyltransferases (PMTs) orchestrate epigenetic modifications through post-translational methylation of various protein substrates including histones. Since dysregulation of this process is widely implicated in many cancers, it is of pertinent interest to screen inhibitors of PMTs, as they offer novel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service