Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09303

Sigma-Aldrich

O-(2-Aminopropyl)-O′-(2-methoxyethyl)polypropylene glycol 500

Synonym(s):

Jeffamine® M-600, Polypropylene glycol 500 mono-2-aminoethyl mono-2-methoxyethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2O[CH(CH3)CH2O]nCH2CH(NH2)CH3
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

SMILES string

COCCO.CC(N)CO.OCCCO

InChI

1S/C3H9NO.2C3H8O2/c1-3(4)2-5;1-5-3-2-4;4-2-1-3-5/h3,5H,2,4H2,1H3;4H,2-3H2,1H3;4-5H,1-3H2

InChI key

XVTJMTQHAYBRLF-UHFFFAOYSA-N

Legal Information

Jeffamine is a registered trademark of Huntsman Petrochemical, LLC

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

264.2 °F - closed cup

flash_point_c

129 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service