Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09283

Sigma-Aldrich

(S)-(+)-1-Amino-2-propanol

purum, ≥98.0% (sum of enantiomers, GC)

Synonym(s):

(+)-Isopropanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH2NH2
CAS Number:
Molecular Weight:
75.11
Beilstein/REAXYS Number:
1718868
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

vapor density

2.6 (vs air)

vapor pressure

<1 mmHg ( 20 °C)

grade

purum

assay

≥98.0% (sum of enantiomers, GC)

optical activity

[α]20/D +18±1°, c = 1.8% in H2O

refractive index

n20/D 1.4437 (lit.)

bp

160 °C (lit.)

mp

24-26 °C (lit.)

density

0.954 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@H](O)CN

InChI

1S/C3H9NO/c1-3(5)2-4/h3,5H,2,4H2,1H3/t3-/m0/s1

InChI key

HXKKHQJGJAFBHI-VKHMYHEASA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

159.8 °F - closed cup

flash_point_c

71 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P C Geiger et al.
Journal of applied physiology (Bethesda, Md. : 1985), 87(5), 1894-1900 (1999-11-24)
The present study examined Ca(2+) sensitivity of diaphragm muscle (Dia(m)) fibers expressing different myosin heavy chain (MHC) isoforms. We hypothesized that Dia(m) fibers expressing the MHC(slow) isoform have greater Ca(2+) sensitivity than fibers expressing fast MHC isoforms and that this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service