09158
1-Propanol
for inorganic trace analysis, ≥99.8%
Synonym(s):
Propyl alcohol
About This Item
Recommended Products
vapor density
2.1 (vs air)
2.1 (vs air)
vapor pressure
10 mmHg ( 147 °C)
10 mmHg
14.9 mmHg ( 20 °C)
assay
≥99.8%
form
liquid
autoignition temp.
700 °F
expl. lim.
13.7 %
8 % (lit.)
impurities
≤0.01% ethanol
≤0.01% methanol
≤0.05% isopropanol
≤0.1% water
≤0.10% water
water
refractive index
n20/D 1.384 (lit.)
bp
97 °C (lit.)
97 °C (lit.)
mp
-127 °C (lit.)
−127 °C (lit.)
density
0.804 g/mL at 25 °C (lit.)
anion traces
bromide (Br-): ≤10 μg/kg
chloride (Cl-): ≤25 μg/kg
fluoride (F-): ≤10 μg/kg
iodide (I-): ≤10 μg/kg
nitrate (NO3-): ≤25 μg/kg
nitrite (NO2-): ≤10 μg/kg
phosphate (PO43-): ≤50 μg/kg
sulfate (SO42-): ≤50 μg/kg
cation traces
Ag: ≤0.5 μg/kg
Al: ≤5 μg/kg
As: ≤1 μg/kg
Au: ≤0.5 μg/kg
Ba: ≤0.5 μg/kg
Be: ≤0.5 μg/kg
Bi: ≤0.5 μg/kg
Ca: ≤30 μg/kg
Cd: ≤0.5 μg/kg
Ce: ≤0.5 μg/kg
Co: ≤0.5 μg/kg
Cr: ≤1 μg/kg
Cs: ≤0.5 μg/kg
Cu: ≤1 μg/kg
Fe: ≤10 μg/kg
Ga: ≤0.5 μg/kg
Ge: ≤0.5 μg/kg
Hg: ≤1 μg/kg
In: ≤0.5 μg/kg
Ir: ≤0.5 μg/kg
K: ≤10 μg/kg
Li: ≤0.5 μg/kg
Mg: ≤10 μg/kg
Mn: ≤10 μg/kg
Mo: ≤0.5 μg/kg
Na: ≤250 μg/kg
Ni: ≤5 μg/kg
Pb: ≤0.5 μg/kg
Pd: ≤0.5 μg/kg
Pt: ≤0.5 μg/kg
Rb: ≤0.5 μg/kg
Rh: ≤0.5 μg/kg
Ru: ≤0.5 μg/kg
Sb: ≤0.5 μg/kg
Se: ≤1 μg/kg
Sn: ≤1 μg/kg
Sr: ≤0.5 μg/kg
Ti: ≤5 μg/kg
Tl: ≤0.5 μg/kg
U: ≤0.5 μg/kg
V: ≤0.5 μg/kg
Zn: ≤10 μg/kg
Zr: ≤0.5 μg/kg
SMILES string
CCCO
InChI
1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3
InChI key
BDERNNFJNOPAEC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
71.6 °F - closed cup
flash_point_c
22 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service