Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09117

Supelco

6-Amino-1-phenalenone

≥97.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9NO
CAS Number:
Molecular Weight:
195.22
Beilstein/REAXYS Number:
2720345
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

derivatization grade ((HPLC))

assay

≥97.0% (HPLC)
≥97.0%

SMILES string

Nc1ccc2C=CC(=O)c3cccc1c23

InChI

1S/C13H9NO/c14-11-6-4-8-5-7-12(15)10-3-1-2-9(11)13(8)10/h1-7H,14H2

InChI key

LNWQVXDBQBAGHD-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service