Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

09105

Supelco

2-Aminoperimidine hydrobromide sesquihydrate

for the determination of SO42-, ≥98.0% (AT)

Synonym(s):

2-Perimidylammonium bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H9N3 · HBr · 1.5H2O
CAS Number:
Molecular Weight:
291.14
Beilstein/REAXYS Number:
3570810
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

Quality Level

assay

≥98.0% (AT)

quality

for the determination of SO42-

technique(s)

UV/Vis spectroscopy: suitable

mp

295-300 °C

SMILES string

Br[H].NC1=Nc2cccc3cccc(N1)c23

InChI

1S/C11H9N3.BrH/c12-11-13-8-5-1-3-7-4-2-6-9(14-11)10(7)8;/h1-6H,(H3,12,13,14);1H

InChI key

HFZUTZWRTHBWPV-UHFFFAOYSA-N

Other Notes

Nephelometric reagent for sulfate determination in trace levels[1][2][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.K. Dasgupta et al.
Mikrochimica Acta, 2, 505-505 (1978)
K.L. Cheng et al. et al.
Handbook of Organic Analytical Reagents null
P.K. Dasgupta et al.
Analytical Chemistry, 50, 1793-1793 (1978)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service