Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08843

Supelco

2-Hydrazinopyridine

≥97.0% (GC)

Synonym(s):

2-Pyridylhydrazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3
CAS Number:
Molecular Weight:
109.13
Beilstein/REAXYS Number:
109984
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

derivatization grade ((HPLC))

Quality Level

assay

≥97.0% (GC)

bp

90-92 °C/1 mmHg (lit.)

mp

41-44 °C (lit.)

storage temp.

2-8°C

SMILES string

NNc1ccccn1

InChI

1S/C5H7N3/c6-8-5-3-1-2-4-7-5/h1-4H,6H2,(H,7,8)

InChI key

NWELCUKYUCBVKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

LC-MS; suitable as derivatisation reagent

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tatsuya Higashi et al.
Steroids, 75(4-5), 338-345 (2010-02-02)
Measurement of steroid levels in saliva has been proposed as a new laboratory tool for characterizing steroid metabolism, but it is not known whether the salivary levels of bile acids can be measured with accuracy and if so, whether such
Yujin Shibayama et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 867(1), 49-56 (2008-04-02)
A sensitive liquid chromatography-electrospray ionization-tandem mass spectrometric (LC-ESI-MS-MS) method for the quantification of 17alpha-hydroxyprogesterone (17OHP) in human saliva has been developed and validated. The saliva was deproteinized with acetonitrile, purified using a Strata-X cartridge, derivatized with a highly proton-affinitive reagent
Tatsuya Higashi et al.
Journal of pharmaceutical and biomedical analysis, 48(1), 177-182 (2008-06-24)
17alpha-hydroxypregnenolone (17OHPreg) has heretofore been considered to be the major cause of the false elevated 17alpha-hydroxyprogesterone (17OHP) value in the immunoassay-based newborn screening for congenital adrenal hyperplasia (CAH). To verify this point, we developed a liquid chromatography-tandem mass spectrometric (LC-MS/MS)
D Hala et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 879(9-10), 591-598 (2011-02-15)
Fathead minnows (Pimephales promelas) comprise a species-of-choice for the hazard assessments of various environmental contaminants, including compounds capable of disrupting endocrine function. Towards this end, the use of liquid chromatography coupled with mass spectrometry (LC-MS) and/or tandem mass spectrometry (MS/MS)
Chemical derivatization to enhance ionization of anabolic steroids in LC-MS for doping-control analysis.
Athanasiadou, I., et al.
Trends in Analytical Chemistry, 42, 137-156 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service