Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08536

Sigma-Aldrich

Sodium thiocyanate 2 M solution

Synonym(s):

Sodium thiocyanate solution, Additive Screening Solution 17/Kit-No 78374, Sodium rhodanide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CNNaS
CAS Number:
Molecular Weight:
81.07
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

[Na+].[N-]=C=S

InChI

1S/CHNS.Na/c2-1-3;/h3H;/q;+1/p-1

InChI key

VGTPCRGMBIAPIM-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1

supp_hazards

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas M Scherer
The journal of physical chemistry. B, 117(8), 2254-2266 (2013-01-22)
The solution thermodynamics and interactions of a reversibly self-associating IgG1 monoclonal antibody have been investigated as a function of cosolute type (NaCl, NaSCN, arginine-HCl) and cosolute concentration over a wide range of protein concentrations (1-235 mg/mL) using static light scattering.
Z G Peng et al.
Colloids and surfaces. B, Biointerfaces, 35(3-4), 169-174 (2004-07-21)
Adsorption and desorption of lysozyme on nano-sized magnetic particles and its conformational change were studied in this work. Adsorption of lysozyme on nano-sized magnetic particles (Fe(3)O(4)) was carried out at different pH. Maximum adsorption of lysozyme (4.65 mg/m2) occurred at
Edyta Glogowska et al.
Blood cells, molecules & diseases, 44(4), 243-251 (2010-03-17)
Historically, the anion transport through the human red cell membrane has been perceived to be mediated by Band 3, in the two-component concept with the large electroneutral anion exchange accompanied by the conductance proper, which dominated the total membrane conductance.
S-F Chin et al.
Molecular pathology : MP, 56(5), 275-279 (2003-09-30)
To describe a robust pretreatment protocol for preparing paraffin wax embedded tissues on tissue microarrays for fluorescence in situ hybridisation (FISH). The newly developed pretreatment protocol described here was compared with the commonly used sodium thiocyanate based protocol and two
Vincent Ball et al.
Langmuir : the ACS journal of surfaces and colloids, 21(9), 4129-4137 (2005-04-20)
In this study, we investigate the buildup of PEI-(PSS-PAH)(n) polyelectrolyte multilayers at pH 7.4 in the presence of either NaCl or NaSCN as a supporting electrolyte. It appears that in the presence of increasing thiocyanate concentrations (from 0.1 to 0.5

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service