Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08453

Sigma-Aldrich

O-(2-tert-Butyloxycarbonylethyl)dodecaethylene glycol

≥95% (oligomer purity)

Synonym(s):

PEG tBu propionate (n=12)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H62O15
Molecular Weight:
674.81
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥95% (oligomer purity)

reaction suitability

reagent type: cross-linking reagent

SMILES string

CC(C)(C)OC(=O)CCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C31H62O15/c1-31(2,3)46-30(33)4-6-34-8-10-36-12-14-38-16-18-40-20-22-42-24-26-44-28-29-45-27-25-43-23-21-41-19-17-39-15-13-37-11-9-35-7-5-32/h32H,4-29H2,1-3H3

InChI key

UOOVFHUCPREVAH-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service