Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08428

Supelco

D-(+)-Fucose

analytical standard

Synonym(s):

6-Deoxy-D-galactose, Rhodeose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12O5
CAS Number:
Molecular Weight:
164.16
Beilstein/REAXYS Number:
1723320
MDL number:
UNSPSC Code:
41116107

grade

analytical standard

Quality Level

assay

≥95% (GC)

optical activity

[α]/D 75±1°, c = 4 in H2O

shelf life

limited shelf life, expiry date on the label

mp

144-145 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H](O)[C@H](O)[C@H](O)[C@@H](O)C=O

InChI

1S/C6H12O5/c1-3(8)5(10)6(11)4(9)2-7/h2-6,8-11H,1H3/t3-,4+,5+,6-/m1/s1

InChI key

PNNNRSAQSRJVSB-DPYQTVNSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

D-(+)-Fucose belonging to the class of sugars (saccharides), is a component of simple glycosides comprising of a few sugar units, generally limited to plant products, microbial, and antibiotic substances.[1]

Application

D-(+)-Fucose may be used as an analytical reference standard for the determination of the analyte in environmental samples using gas chromatography coupled to mass spectrometry (GC–MS) technique.[2]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Challenges in the enantioanalysis of fucose using stochastic and potentiometric microsensors
Moldoveanu I, et al.
Sensing and Bio-Sensing Research, 1, 1-7 (2014)
Development of Gradient Retention Model in Ion Chromatography. Part II: Artificial Intelligence QSRR Approach
Ukic S, et al.
Chromatographia, 77, 997-1007 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service