Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08357

Sigma-Aldrich

Glutathione reduced diethyl ester

BioChemika

Synonym(s):

GSH diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H25N3O6S
CAS Number:
Molecular Weight:
363.43
UNSPSC Code:
12352202
Pricing and availability is not currently available.

product line

BioChemika

storage temp.

2-8°C

InChI

1S/C14H25N3O6S/c1-3-22-12(19)7-16-13(20)10(8-24)17-11(18)6-5-9(15)14(21)23-4-2/h9-10,24H,3-8,15H2,1-2H3,(H,16,20)(H,17,18)/t9-,10-/m0/s1

InChI key

IYXATRBAHYMOQL-UWVGGRQHSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Detection and affinity purification of oxidant-sensitive proteins using biotinylated glutathione ethyl ester.
Daniel M Sullivan et al.
Methods in enzymology, 353, 101-113 (2002-06-25)
P J Thornalley et al.
Biochemical pharmacology, 47(2), 418-420 (1994-01-20)
S-p-Bromobenzylglutathione diethyl ester induced toxicity in the malarial parasite Plasmodium falciparum in infected human red blood cells in culture. The median inhibitory concentration, IC50, was 4.77 +/- 0.12 microM (N = 10) for incorporation of [3H]hypoxanthine in nucleotide synthesis and
E J Levy et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(19), 9171-9175 (1993-10-01)
Glutathione monoesters in which the carboxyl group of the glycine residue is esterified were previously found, in contrast to glutathione itself, to be effectively transported into various types of cells and to be converted intracellularly into glutathione. Glutathione monoesters are
Akiko Fukumoto et al.
Investigative ophthalmology & visual science, 51(5), 2450-2458 (2010-01-01)
Compounds regulating intracellular thiol redox status, such as N,N-diacetyl-L-cystine dimethylester (NM(2)), were shown to prolong corneal graft survival in a penetrating keratoplasty (PKP) model. However, the effect of NM(2) on hemangiogenesis and lymphangiogenesis has not been investigated. The effect of
Agata Klejman et al.
Neurochemistry international, 47(1-2), 51-57 (2005-06-30)
The occurrence, nature and prevention of ammonia-induced cell death were assayed in cultured primary cortical neurons from newborn rats. Treatment with 1-10 mM ammonium chloride for 24 or 48 h, dose-dependently decreased neuronal survival (MTT assay) and GSH/GSSG ratio in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service