Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08091

Sigma-Aldrich

SDS Gel Preparation Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500

impurities

acrylamide
bisacrylamide

Quality Level

storage temp.

2-8°C

Application

Ready to use solutions for ~50 SDS-PAGE gels (gel size 8x8 cm, 15 ml)

Components

Components:
250 mL Separation Gel Buffer, 3× Concentrate (Cat. No. 14946)
100 mL Stacking Gel Buffer, 5× Concentrate (Cat. No. 19505)
250 mL 30% Acrylamide/0.8% Bisacrylamide Solution (Cat. No. 06867)
3x 250 mL Buffer Concentrate, 10× Concentrate (Cat. No. 14393)
30 mL TEMED 10% (Cat. No. 07654)
30 mL Incubation Buffer Concentrate
10 g Ammonium Persulfate (APS)

related product

Product No.
Description
Pricing

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Liq. 3 - Repr. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 1 Oral - STOT SE 3

target_organs

Peripheral nervous system, Respiratory system

Storage Class

5.1B - Oxidizing hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura P Frazão et al.
Biomolecules, 10(9) (2020-08-23)
Although some placenta-derived products are already used for tissue regeneration, the human chorion membrane (HCM) alone has been poorly explored. In fact, just one study uses decellularized HCM (dHCM) with native tissue architecture (i.e., without extracellular matrix (ECM) suspension creation)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service