Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

08051

Sigma-Aldrich

2-Aminohexadecanoic acid

technical, ≥95% (NT)

Synonym(s):

2-Aminopalmitic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H33NO2
CAS Number:
Molecular Weight:
271.44
Beilstein/REAXYS Number:
1785113
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

assay

≥95% (NT)

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

color

white to faint beige

mp

~220 °C (dec.)

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

CCCCCCCCCCCCCCC(N)C(O)=O

InChI

1S/C16H33NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(17)16(18)19/h15H,2-14,17H2,1H3,(H,18,19)

InChI key

XELWBYCKQCNAGY-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicolas Basse et al.
Bioorganic & medicinal chemistry letters, 16(12), 3277-3281 (2006-04-25)
Proteasomes are responsible for the cytoplasmic turnover of the vast majority of proteins including regulatory proteins. We have synthesized lipopeptides a new class of non-covalent inhibitors of the 20S proteasome and assayed their inhibitory capacities. Their ability to inhibit at

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service