Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07900

Sigma-Aldrich

N-Phenyl-o-phenylenediamine

purum, ≥98.0% (NT)

Synonym(s):

2-Aminodiphenylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5NHC6H4NH2
CAS Number:
Molecular Weight:
184.24
Beilstein/REAXYS Number:
908984
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

assay

≥98.0% (NT)

mp

77-80 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1ccccc1Nc2ccccc2

InChI

1S/C12H12N2/c13-11-8-4-5-9-12(11)14-10-6-2-1-3-7-10/h1-9,14H,13H2

InChI key

NFCPRRWCTNLGSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriele Manzella et al.
Nature communications, 11(1), 4629-4629 (2020-09-17)
Cancer therapy is currently shifting from broadly used cytotoxic drugs to patient-specific precision therapies. Druggable driver oncogenes, identified by molecular analyses, are present in only a subset of patients. Functional profiling of primary tumor cells could circumvent these limitations, but

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service