Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07784

Sigma-Aldrich

1-Ethyl-3-methylimidazolium ethyl sulfate

for catalysis, ≥98.5% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16N2O4S
CAS Number:
Molecular Weight:
236.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

≥98.5% (T)

impurities

≤200 ppm water

refractive index

n20/D 1.481

SMILES string

CCOS([O-])(=O)=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H6O4S/c1-3-8-5-4-7(2)6-8;1-2-6-7(3,4)5/h4-6H,3H2,1-2H3;2H2,1H3,(H,3,4,5)/q+1;/p-1

InChI key

VRFOKYHDLYBVAL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

310.1 °F

flash_point_c

154.5 °C

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fernando Benito-Lopez et al.
Lab on a chip, 14(18), 3530-3538 (2014-07-16)
This paper reports for the first time the use of a cross-linked poly(N-isopropylacrylamide) ionogel encapsulating the ionic liquid 1-ethyl-3-methylimidazolium ethyl sulphate as a thermoresponsive and modular microfluidic valve. The ionogel presents superior actuation behaviour to its equivalent hydrogel. Ionogel swelling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service