Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07676

Sigma-Aldrich

Lipase, Thermomyces lanuginosa, CLEA

≥25 U/mg

Synonym(s):

Lipase, Thermomyces lanuginosa, cross-linked enzyme aggregate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.22

form

powder

Quality Level

specific activity

≥25 U/mg

storage temp.

2-8°C

Unit Definition

1 U corresponds to the amount of enzyme which sets free1 μmol butyric acid per minute at pH 7.5 and 40°C (glyceryl tributyrate, Cat. No. 91010, as substrate)

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

One-step lipase-catalysed preparation of eslicarbazepine.
El-Behairy MF & Sundby E.
Royal Society of Chemistry Advances, 6(101), 98730-98736 (2016)
Effect of different parameters on the hydrolytic activity of cross-linked enzyme aggregates (CLEAs) of lipase from Thermomyces lanuginosa.
Torres MG, et al.
Biochemical Engineering Journal, 72, 18-23 (2013)
Biocatalyst mediated regio-and stereo-selective hydroxylation and epoxidation of (Z)-?-santalol.
Daramwar PP, et al.
Organic & Biomolecular Chemistry, 12(7), 1048-1051 (2014)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service