Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07527

Supelco

O-Desmethylindomethacin

analytical standard

Synonym(s):

1-(4-Chlorobenzoyl)-5-hydroxy-2-methyl-3-indoleacetic acid, 5-Hydroxyindomethacin, Demethylindomethacin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14ClNO4
CAS Number:
Molecular Weight:
343.76
Beilstein/REAXYS Number:
443408
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.76

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC1=C(CC(O)=O)C2=CC(O)=CC=C2N1C(C3=CC=C(Cl)C=C3)=O

InChI

1S/C18H14ClNO4/c1-10-14(9-17(22)23)15-8-13(21)6-7-16(15)20(10)18(24)11-2-4-12(19)5-3-11/h2-8,21H,9H2,1H3,(H,22,23)

InChI key

KMLNWQPYFBIALN-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service