Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07501

Supelco

α-Terpineol-(propyl methyl-d3) solution

1 mg/mL in isooctane, analytical standard

Synonym(s):

α,4-Dimethyl-α-(methyl-d3)-3-cyclohexene-1-methanol, α-Terpineol-d3 solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D3H15O
CAS Number:
Molecular Weight:
157.27
UNSPSC Code:
41116107

grade

analytical standard

assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

concentration

1 mg/mL in isooctane

refractive index

n20/D 1.391-1.393

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

CC1=CCC(C(O)(C)C([2H])([2H])[2H])CC1

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

10.4 °F - closed cup

flash_point_c

-12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service