Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07448

Epoxide Hydrolase, Rhodococcus erythropolis, recombinant from E. coli

Synonym(s):

Hydratase, epoxide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

recombinant

expressed in E. coli

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magdalena E Potok et al.
Proceedings of the National Academy of Sciences of the United States of America, 119(3) (2022-01-15)
ARABIDOPSIS TRITHORAX-RELATED PROTEIN 5 (ATXR5) AND ATXR6 are required for the deposition of H3K27me1 and for maintaining genomic stability in Arabidopsis Reduction of ATXR5/6 activity results in activation of DNA damage response genes, along with tissue-specific derepression of transposable elements
Edwin R Smith et al.
Molecular and cellular biology, 28(3), 1041-1046 (2007-11-28)
Histone H3 methylation at Lys27 (H3K27 methylation) is a hallmark of silent chromatin, while H3K4 methylation is associated with active chromatin regions. Here we report that a Drosophila JmjC family member, dUTX, specifically demethylates di- and trimethylated but not monomethylated
G Foltz et al.
Oncogene, 28(29), 2667-2677 (2009-05-26)
Epigenetic inactivation of tumor suppressor genes is a common feature in human cancer. Promoter hypermethylation and histone deacetylation are reversible epigenetic mechanisms associated with transcriptional regulation. DNA methyltransferases (DNMT1 and DNMT3b) regulate and maintain promoter methylation and are overexpressed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service