Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07389

Supelco

Benzyl 4-hydroxybenzoate

analytical standard

Synonym(s):

4-Hydroxybenzoic acid benzyl ester, Benzyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4CO2CH2C6H5
CAS Number:
Molecular Weight:
228.24
Beilstein/REAXYS Number:
2115995
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

mp

109-112 °C (lit.)

application(s)

environmental

format

neat

SMILES string

Oc1ccc(cc1)C(=O)OCc2ccccc2

InChI

1S/C14H12O3/c15-13-8-6-12(7-9-13)14(16)17-10-11-4-2-1-3-5-11/h1-9,15H,10H2

InChI key

MOZDKDIOPSPTBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl 4-hydroxybenzoate is an ester of para-hydroxy benzoic acid. It is used as a preservative, colorless dye and color developing agent in cosmetics and personal care products.[1]

Application

Benzyl 4-hydroxybenzoate may be used as an analytical standard for the determination of the analyte in cosmetics, personal care products and pharmaceutical formulations by chromatography[2][3] and spectrometry[4] techniques.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Preservatives in Cosmetics and Personal Care Products by LC-MS-MS
Myers EA, et al.
LCGC North America, 33, 16-22 (2015)
Determination of parabens in pharmaceutical formulations by solid-phase microextraction-ion mobility spectrometry
Lokhnauth JK and Snow NH
Analytical Chemistry, 77(18), 5938-5946 (2005)
Validation of HPLC analysis of 2-phenoxyethanol, 1-phenoxypropan-2-ol, methyl, ethyl, propyl, butyl and benzyl 4-hydroxybenzoate (parabens) in cosmetic products, with emphasis on decision limit and detection capability.
Borremans M, et al.
Chromatographia, 59(1-2), 47-53 (2004)
Hung-Ming Yang et al.
Ultrasonics sonochemistry, 21(1), 395-400 (2013-08-27)
The catalytic esterification of sodium 4-hydroxybenzoate with benzyl bromide by ultrasound-assisted solid-liquid phase-transfer catalysis (U-SLPTC) was investigated using the novel dual-site phase-transfer catalyst 4,4'-bis(tributylammoniomethyl)-1,1'-biphenyl dichloride (BTBAMBC), which was synthesized from the reaction of 4,4'-bis(chloromethyl)-1,1'-biphenyl and tributylamine. Without catalyst and in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service