Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07362

Supelco

Fenthion oxon sulfoxide-(S-methyl-d3)

PESTANAL®, analytical standard

Synonym(s):

Dimethyl-[3-methyl-4-(methyl-d3-sulfinyl)phenyl]-phosphat, Fenoxon sulfoxide-(S-methyl-d3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D3H12O5PS
Molecular Weight:
281.28
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=P(OC)(OC)OC1=CC(C)=C(S(C([2H])([2H])[2H])=O)C=C1

General description

Fenthion oxon sulfoxide-(S-methyl-d3) is an isotope-labeled analog of fenthion oxon,[1] a metabolite of the organophosphorus pesticide fenthion,[2] wherein the S-methyl protons are replaced by deuterium.

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of fenthion and temephos and their transformation products in water by high-performance liquid chromatography with diode array detection and atmospheric pressure chemical ionization mass spectrometric detection
Lacorte S, et al.
Journal of Chromatography A, 777(1), 99-114 (1997)
Antiandrogenic activity and metabolism of the organophosphorus pesticide fenthion and related compounds.
Kitamura S, et al.
Environmental Health Perspectives, 111(4), 503-508 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service