Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07303

Supelco

(2RS)-2-Ethyl-2-methylpentanoic acid

pharmaceutical impurity standard

Synonym(s):

Valproic acid impurity K (PhEur), 2-Ethyl-2-methylvaleric acid, NSC 466

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21
Beilstein/REAXYS Number:
1721454
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Quality Level

assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

application(s)

clinical testing

format

neat

SMILES string

OC(C(C)(CC)CCC)=O

InChI

1S/C8H16O2/c1-4-6-8(3,5-2)7(9)10/h4-6H2,1-3H3,(H,9,10)

InChI key

WUWPVNVBYOKSSZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service