Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

07166

3-[4-(Bromomethyl)phenyl]-7-(diethylamino)coumarin

Synonym(s):

3-(4-(Bromomethyl)phenyl)-7-(diethylamino)-2H-1-benzopyran-2-one, MPAC-Br

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20BrNO2
CAS Number:
Molecular Weight:
386.28
MDL number:
UNSPSC Code:
41100000
PubChem Substance ID:

storage temp.

2-8°C

SMILES string

CCN(CC)c1ccc2C=C(C(=O)Oc2c1)c3ccc(CBr)cc3

InChI

1S/C20H20BrNO2/c1-3-22(4-2)17-10-9-16-11-18(20(23)24-19(16)12-17)15-7-5-14(13-21)6-8-15/h5-12H,3-4,13H2,1-2H3

InChI key

WTCCANMBFQWZMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service