Skip to Content
MilliporeSigma

07095

4-Aminobenzyl alcohol

purum, ≥97.0% (T)

Synonym(s):

4-(Hydroxymethyl)aniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2NC6H4CH2OH
CAS Number:
Molecular Weight:
123.15
EC Number:
210-767-5
UNSPSC Code:
12352100
PubChem Substance ID:
Beilstein/REAXYS Number:
2078680
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

purum

assay

≥97.0% (T)

mp

60-65 °C

storage temp.

2-8°C

SMILES string

Nc1ccc(CO)cc1

InChI

1S/C7H9NO/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5,8H2

InChI key

AXKGIPZJYUNAIW-UHFFFAOYSA-N



Still not finding the right product?

Explore all of our products under 4-Aminobenzyl alcohol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Franciscus M H de Groot et al.
Molecular cancer therapeutics, 1(11), 901-911 (2002-12-17)
The design, synthesis, and initial biological evaluation of a doxorubicin prodrug that contains a dual tumor specific moiety, which allows enhanced tumor recognition potential, is reported. Both a tumor-specific recognition site and a tumor selective enzymatic activation sequence are incorporated
C Elvira et al.
Biomaterials, 23(9), 1955-1966 (2002-05-09)
The design and preparation of novel biodegradable hydrogels developed by the free radical polymerization of acrylamide and acrylic acid, and some formulations with bis-acrylamide, in the presence of a corn starch/ethylene-co-vinyl alcohol copolymer blend (SEVA-C), is reported. The redox system