Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06997

Supelco

(R)-2-Methyl-3-ureidopropionic acid

Synonym(s):

(2R)-3-[(Aminocarbonyl)amino]-2-methylpropanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10N2O3
CAS Number:
Molecular Weight:
146.14
UNSPSC Code:
12352200
NACRES:
NA.24

assay

≥85% (HPLC)

Quality Level

optical activity

[α]/D -19±2°, c = 0.1 in ethanol

format

neat

storage temp.

2-8°C

SMILES string

O=C(O[H])[C@@](C([H])([H])[H])([H])C([H])([H])N([H])C(N([H])[H])=O

Biochem/physiol Actions

This metabolite is increased in the urine of patients with beta-ureidopropionase deficiency and can be used to predict patient′s individual phenotypes of enzyme deficiencies in pyrimidine metabolism when associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Urinary metabonomics of stomach cancer assessed by rapid resolution liquid chromatography/time-of-flight mass spectrometry
Zhang, Y., et al.
Chinese Medical Journal (English Edition), 126, 1930-1933 (2013)
NMR-based urinalysis for rapid diagnosis of ?-ureido- propionase deficiency in a patient with Dravet syndrome.
Lam, C.W., et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 440, 201-204 (2015)
Morimasa Ohse et al.
Journal of mass spectrometry : JMS, 37(9), 954-962 (2002-09-25)
Dihydropyrimidine dehydrogenase (DHPDase), dihydropyrimidinase (DHPase) and beta-ureidopropionase (betaUPase) are the enzymes that catalyze the first, second, and third steps of the degradation of pyrimidines, respectively. beta-Ureidopropionate (betaUP) and beta-ureidoisobutyrate (betaUIB) are increased in the urine of patients with betaUPase deficiency.
Ute Hofmann et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 791(1-2), 371-380 (2003-06-12)
Enzyme deficiencies in pyrimidine metabolism are associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil. To assess whether urinary levels of pyrimidines and their metabolites can be used for predicting patients' individual phenotype, a new gas chromatographic-tandem
André B P van Kuilenburg et al.
Biochimica et biophysica acta, 1822(7), 1096-1108 (2012-04-25)
ß-ureidopropionase is the third enzyme of the pyrimidine degradation pathway and catalyzes the conversion of N-carbamyl-ß-alanine and N-carbamyl-ß-aminoisobutyric acid to ß-alanine and ß-aminoisobutyric acid, ammonia and CO(2). To date, only five genetically confirmed patients with a complete ß-ureidopropionase deficiency have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service