Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06963

Supelco

4-(Diethylamino)benzhydrazide

≥98.0% (HPLC)

Synonym(s):

4-(Diethylamino)benzoic acid hydrazide, 4-(Diethylamino)benzoylhydrazide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17N3O
CAS Number:
Molecular Weight:
207.27
Beilstein/REAXYS Number:
2731205
MDL number:
UNSPSC Code:
41121800
PubChem Substance ID:
NACRES:
NB.21

grade

derivatization grade ((HPLC))

Quality Level

assay

≥98.0% (HPLC)

storage temp.

2-8°C

SMILES string

O=C(NN)C1=CC=C(N(CC)CC)C=C1

InChI

1S/C11H17N3O/c1-3-14(4-2)10-7-5-9(6-8-10)11(15)13-12/h5-8H,3-4,12H2,1-2H3,(H,13,15)

InChI key

PUNSOBLTEWFAMJ-UHFFFAOYSA-N

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Skin Sens. 1

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshio Suzuki et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 20(3), 475-482 (2004-04-08)
Novel labeling reagents, called MS probes, which possess a positively charged quaternary amine moiety and can transform a neutral analyte into a charged compound by simply mixing with the analyte and allowing the mixture to stand from several minutes to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service