Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06708

Supelco

Neoeriocitrin

analytical standard

Synonym(s):

(S)-3′,4′,5,7-Tetrahydroxyflavanone-7-[2-O-(α-L-rhamnopyranosyl)-β-D-glucopyranoside], Eriodictyol 7-neohesperidoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H32O15
CAS Number:
Molecular Weight:
596.53
Beilstein/REAXYS Number:
1696764
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

assay

≥97.0% (HPLC)

form

powder

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](O)[C@@H](CO)O[C@H]2Oc3cc(O)c4C(=O)C[C@H](Oc4c3)c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H32O15/c1-9-20(33)22(35)24(37)26(38-9)42-25-23(36)21(34)18(8-28)41-27(25)39-11-5-14(31)19-15(32)7-16(40-17(19)6-11)10-2-3-12(29)13(30)4-10/h2-6,9,16,18,20-31,33-37H,7-8H2,1H3/t9-,16-,18+,20-,21+,22+,23-,24+,25+,26-,27+/m0/s1

InChI key

OBKKEZLIABHSGY-DOYQYKRZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Neoeriocitrin was one of several flavonoids that completely blocked the okadaic acid-induced inhibition of autophagy in cultured mouse hepatocytes. It is suggested as tool in the study of intracellular protein phosphorylation and as a potential protectant against pathological hyperphosphorylations.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shufang Wang et al.
Journal of chromatography. A, 1348, 105-124 (2014-05-13)
The anti-inflammatory constituents of Ju-Zhi-Jiang-Tang (JZJT), a formula used for thousands of years in China, were identified by LC-MS and pharmacological activity evaluation. In this study, the whole extract of formula was separated into multiple components to facilitate the analytical

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service