Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06683

Supelco

Maize GMO Standard ERM-BF414

ERM®, certified reference material, Set for 0%, 0.1%, 0.5%, 1%, 1.7%, 4.3% GA21

Synonym(s):

GMO Standard ERM-BF414, Maize GA21

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

grade

certified reference material

agency

ERM®

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Legal Information

ERM is a registered trademark of European Commission

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service