Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06681

Supelco

Epoxidized linseed oil

analytical standard

Synonym(s):

ELO, Linseed oil, epoxidized

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

impurities

~9% epoxides

refractive index

n20/D 1.477

density

1.034 g/mL at 20 °C (lit.)

format

neat

Looking for similar products? Visit Product Comparison Guide

Other Notes

Qualitative standard for the determination of ELO in foods with HPLC-MS or GC-MS. ELO is currently still employed as plasticizer and HCl scavenger in foamed PVC used in cap linings

Storage Class

10 - Combustible liquids

wgk_germany

awg

flash_point_f

590.0 °F - closed cup

flash_point_c

310.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service