06244
Arjunic acid
analytical standard
Synonym(s):
(2α,3β,19α)-2,3,19-Trihydroxyolean-12-en-28-oic acid, Arjuntriterpenic acid
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥88.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
application(s)
food and beverages
format
neat
SMILES string
CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]2[C@@H]1O)C(O)=O
InChI
1S/C30H48O5/c1-25(2)12-14-30(24(34)35)15-13-28(6)17(21(30)23(25)33)8-9-20-27(5)16-18(31)22(32)26(3,4)19(27)10-11-29(20,28)7/h8,18-23,31-33H,9-16H2,1-7H3,(H,34,35)/t18-,19+,20-,21-,22+,23+,27+,28-,29-,30+/m1/s1
InChI key
XJMYUPJDAFKICJ-YZTGESMESA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service