06224
2-Isobutyl-3-methoxypyrazine
analytical standard
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥99% (GC)
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.488-1.492
n20/D 1.49 (lit.)
density
0.99 g/mL at 25 °C (lit.)
suitability
conforms to structure for Proton NMR spectrum
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
SMILES string
COc1nccnc1CC(C)C
InChI
1S/C9H14N2O/c1-7(2)6-8-9(12-3)11-5-4-10-8/h4-5,7H,6H2,1-3H3
InChI key
UXFSPRAGHGMRSQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
176.0 °F - closed cup
flash_point_c
80 °C - closed cup
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Separation of 2-Isopropyl-3-methoxypyrazine 20 ppt; 2-Isobutyl-3-methoxypyrazine; 2-Methylisoborneol; (±)-Geosmin 10 ppt
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service