Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

06011501

Mint oil

secondary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

matrix material

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Phytochemicals: (-)-Menthol, (-)-Menthone (quantitative); Limonene, Menthyl acetate, Isopulegol (qualitative)

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

155.7 °F

flash_point_c

68.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service