grade
analytical standard
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
HWI
refractive index
n20/D 1.4665 (lit.)
n20/D 1.4675 (lit.)
bp
159 °C (lit.)
density
0.877 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
matrix material
Looking for similar products? Visit Product Comparison Guide
General description
Exact content by quantitative NMR can be found on the certificate.
signalword
Danger
hcodes
Hazard Classifications
Asp. Tox. 1 - Flam. Liq. 3 - Skin Irrit. 2
Storage Class
3 - Flammable liquids
wgk_germany
WGK 3
flash_point_f
132.8 °F - closed cup
flash_point_c
56 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service