Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05630590

(±)-β-Citronellol

primary reference standard

Synonym(s):

3,7-Dimethyl-6-octen-1-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CH2OH
CAS Number:
Molecular Weight:
156.27
Beilstein/REAXYS Number:
1721507
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.24

grade

primary reference standard

vapor density

5.4 (vs air)

vapor pressure

~0.02 mmHg ( 25 °C)

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.456 (lit.)

bp

222 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

food and beverages

Storage temp.

−20°C

SMILES string

CC(CCO)CC\C=C(\C)C

InChI

1S/C10H20O/c1-9(2)5-4-6-10(3)7-8-11/h5,10-11H,4,6-8H2,1-3H3

Inchi Key

QMVPMAAFGQKVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

224.6 °F - closed cup

flash_point_c

107 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service