Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05598

Millipore

Feeley Gorman Agar

suitable for microbiology

Synonym(s):

FG Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106200

shelf life

limited shelf life, expiry date on the label

composition

agar, 17.0 g/L
beef extract, 3.0 g/L
casein acid hydrolysate, 17.5 g/L
L-cysteine hydrochloride, 0.4 g/L
ferric pyrophosphate, soluble, 0.25 g/L
starch, 1.5 g/L

final pH

6.9±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages

microbiology

suitability

nonselective for Legionella spp.

General description

Feeley Gorman Agar / Broth is recommended for isolation and presumptive identification of Legionella species[1].

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G A Hébert et al.
Annals of internal medicine, 92(1), 53-54 (1980-01-01)
The "Pittsburgh pneumonia agent," isolated by Pasculle and co-workers from human lung tissue, has been cultured on artificial media and characterized. The "Pittsburgh" bacterium and the TATLOCK and HEBA bacteria have identical cultural, biochemical, and antigenic characteristics. They also have
J C Feeley et al.
Journal of clinical microbiology, 8(3), 320-325 (1978-09-01)
Yolk sac suspensions infected with the Legionnaires disease bacterium (LDB) were plated onto 17 different bacteriological agar media. The LDB grew only on Mueller-Hinton agar supplemented with 1% Iso Vitale X and 1% hemoglobin (MH-IH). This medium was subsequently analyzed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service