Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05149

Supelco

Thiogeraniol

analytical standard

Synonym(s):

3,7-Dimethyl-2,6-octadiene-1-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18S
CAS Number:
Molecular Weight:
170.31
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥85.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.501-1.503

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

shipped in

dry ice

storage temp.

−20°C

SMILES string

C\C(C)=C\CC\C(C)=C\CS

InChI

1S/C10H18S/c1-9(2)5-4-6-10(3)7-8-11/h5,7,11H,4,6,8H2,1-3H3/b10-7+

InChI key

FACAUSJJVBMWLV-JXMROGBWSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service