Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

05098

Supelco

Cyanidin 3-O-α-L-arabinoside chloride

analytical standard

Synonym(s):

Cyanidin 3-α-L-arabinopyranoside, Cyanidin 3-α-L-arabinoside, Cyanidin 3-arabinoside chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClO10
CAS Number:
Molecular Weight:
454.81
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤5.0% water

application(s)

food and beverages

format

neat

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC1=CC(O)=C(C=C(O[C@@H]2OC[C@H](O)[C@H](O)[C@H]2O)C(C3=CC=C(O)C(O)=C3)=[O+]4)C4=C1.[Cl-]

InChI

1S/C20H18O10.ClH/c21-9-4-12(23)10-6-16(30-20-18(27)17(26)14(25)7-28-20)19(29-15(10)5-9)8-1-2-11(22)13(24)3-8;/h1-6,14,17-18,20,25-27H,7H2,(H3-,21,22,23,24);1H/t14-,17-,18+,20-;/m0./s1

InChI key

ORTBMTXABUAMJS-HAHUOHMJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service