04969
(S)-(−)-Perillaldehyde
analytical standard
Synonym(s):
(−)-Perillaaldehyde, (S)-4-Isopropenyl-cyclohexene-1-carboxaldehyde, (S)-p-Mentha-1,8-dien-7-al
About This Item
Recommended Products
grade
analytical standard
Quality Level
assay
≥97% (GC)
optical activity
[α]/D -125±5°, c = 10 in ethanol
shelf life
limited shelf life, expiry date on the label
refractive index
n20/D 1.508 (lit.)
bp
104-105 °C/10 mmHg (lit.)
density
0.965 g/mL at 25 °C (lit.)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
format
neat
storage temp.
2-8°C
SMILES string
CC(=C)[C@H]1CCC(C=O)=CC1
InChI
1S/C10H14O/c1-8(2)10-5-3-9(7-11)4-6-10/h3,7,10H,1,4-6H2,2H3/t10-/m1/s1
InChI key
RUMOYJJNUMEFDD-SNVBAGLBSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
204.8 °F - closed cup
flash_point_c
96 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
A throughput HS-SPME-GC-MS method to analyze volatile compounds in Perilla leaves at low extraction temperatures using four different SPME fibers.
Protocols
-β-Farnesene; α-Huµlene; Germacrene D; (+)-Valencene; Bicyclogermacrene; (+)-δ-Cadinene
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service