Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04895

NIR 6a

Synonym(s):

1-(3-Aminopropyl)-4,5-benzo-11-chloro-1′-(4-sulfobutyl)-3,3,3′,3′-tetramethyl-10,12-trimethylenindotricarbocyanine bromide hydrobromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H48BrClN3NaO3S · HBr
CAS Number:
Molecular Weight:
882.16
UNSPSC Code:
41100000
Pricing and availability is not currently available.

storage temp.

2-8°C

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service