Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04885

Supelco

L-Ascorbic acid 6-stearate

analytical standard

Synonym(s):

6-O-Stearyl-L-ascorbic acid, L-Ascorbyl 6-octadecanoate, Ascorbyl stearate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H42O7
CAS Number:
Molecular Weight:
442.59
EC Number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

InChI

1S/C24H42O7/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-20(26)30-18-19(25)23-21(27)22(28)24(29)31-23/h19,23,25,28-29H,2-18H2,1H3

InChI key

NRHBEGWPKVFGRH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Ascorbic acid 6-stearate is a lipophilic fatty acid ester of ascorbic acid.[1]
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

L-Ascorbic acid 6-stearate (ASC-S) may be used as a standard in determining the apoptotic activity of ASC-S through the intrinsic mitochondrial pathway in HeLa cell lines, derived from cervical cancer cells.[2] It may also be used as an analytical standard to investigate its interaction with bovine serum albumin using surface plasmon resonance (SPR).[3]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetic and thermodynamic studies of bovine serum albumin interaction with ascorbyl palmitate and ascorbyl stearate food additives using surface plasmon resonance
Fathi F, et al.
Food Chemistry, 246(6B), 228-232 (2018)
Ascorbyl stearate promotes apoptosis through intrinsic mitochondrial pathway in HeLa cancer cells
Mane DS, et al.
Anticancer Research, 36(12), 6409-6417 (2016)
P53 enhances ascorbyl stearate-induced G2/M arrest of human ovarian cancer cells
Naidu AK, et al.
Anticancer Research, 27(6B), 3927-3934 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service