Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04649

Supelco

3-Hexanol

analytical standard

Synonym(s):

Ethyl propyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH(OH)CH2CH3
CAS Number:
Molecular Weight:
102.17
Beilstein/REAXYS Number:
1718964
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.415 (lit.)
n20/D 1.416

bp

134-135 °C (lit.)

density

0.820 g/mL at 20 °C
0.819 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCC(O)CC

InChI

1S/C6H14O/c1-3-5-6(7)4-2/h6-7H,3-5H2,1-2H3

InChI key

ZOCHHNOQQHDWHG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

113.0 °F - closed cup

flash_point_c

45 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service