Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04590595

Menthyl acetate

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

refractive index

n20/D 1.447 (lit.)

bp

228-229 °C (lit.)

density

0.922 g/mL at 25 °C (lit.)

application(s)

food and beverages

storage temp.

−20°C

SMILES string

CC(C)C1CCC(C)CC1OC(C)=O

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3

InChI key

XHXUANMFYXWVNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

pictograms

Environment

hcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

197.6 °F - closed cup

flash_point_c

92 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service