Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04527

Sigma-Aldrich

Z-Nle-OH

≥98.0%

Synonym(s):

Z-L-Norleucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H19NO4
CAS Number:
Molecular Weight:
265.30
Beilstein/REAXYS Number:
3210973
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Quality Level

assay

≥98.0%

form

powder

reaction suitability

reaction type: solution phase peptide synthesis

technique(s)

ligand binding assay: suitable

color

white

application(s)

peptide synthesis

SMILES string

CCCC[C@H](NC(=O)OCc1ccccc1)C(O)=O

InChI

1S/C14H19NO4/c1-2-3-9-12(13(16)17)15-14(18)19-10-11-7-5-4-6-8-11/h4-8,12H,2-3,9-10H2,1H3,(H,15,18)(H,16,17)/t12-/m0/s1

InChI key

NMYWMOZOCYAHNC-LBPRGKRZSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tianliang He et al.
Oncogene (2018-06-22)
The antiviral metabolites from bacterial stress response to bacteriophage infection can maintain homeostasis of host cells, while metabolism disorder is a remarkable characteristic of tumorigenesis. In the aspect of metabolic homeostasis, therefore, the antiviral homeostasis-maintaining metabolites of bacteria may possess

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service