Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04491

Supelco

2-Myristoylglycerol

analytical standard

Synonym(s):

β-Monomyristin, 2-Monomyristin, 2-Tetradecanoylglycerol, Myristic acid β-monoglyceride, Tetradecanoic acid 2-hydroxy-1-(hydroxymethyl)ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H34O4
CAS Number:
Molecular Weight:
302.45
Beilstein/REAXYS Number:
1792657
UNSPSC Code:
85151701
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

analytical standard

assay

≥95.0% (HPLC)

form

powder or crystals

shelf life

limited shelf life, expiry date on the label

functional group

ester

storage temp.

−70°C

SMILES string

CCCCCCCCCCCCCC(=O)OC(CO)CO

InChI

1S/C17H34O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-17(20)21-16(14-18)15-19/h16,18-19H,2-15H2,1H3

InChI key

TVIMZSOUQXNWHO-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service