About This Item
Recommended Products
assay
≥99.0% (NT)
impurities
≤1.0% water
mp
≥40 °C
SMILES string
[O-][N+]([O-])=O.CCn1cc[n+](C)c1
InChI
1S/C6H11N2.NO3/c1-3-8-5-4-7(2)6-8;2-1(3)4/h4-6H,3H2,1-2H3;/q+1;-1
InChI key
JDOJFSVGXRJFLL-UHFFFAOYSA-N
Other Notes
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
5.1B - Oxidizing hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service