Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

04311

Supelco

Maleic hydrazide-13C4

PESTANAL®, analytical standard

Synonym(s):

1,2-Dihydro-3,6-pyridazine-13C4-dione, 3,6-Dihydroxypyridazine-13C4, 3,6-Pyridazine-13C4-diol, Maleic acid hydrazide-13C4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C4H4N2O2
Molecular Weight:
116.06
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture

format

neat

General description

Maleic hydrazide-13C4 is an isotope-labeled analog of maleic hydrazide, a plant growth inhibitor with herbicidal activity, wherein the pyridazine ring protons are replaced by deuterium.[1][2]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The major advantage of using this technique is that the isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analogs, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

572.0 °F

flash_point_c

300 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of matrix solid-phase dispersion to the propham and maleic hydrazide determination in potatoes by differential pulse voltammetry and HPLC
Arribas AS, et al.
Talanta, 71(1), 430-436 (2007)
Determination of maleic hydrazide in potatoes and onions by capillary electrophoresis
Kubilius DT and Bushway RJ
Journal of Agricultural and Food Chemistry, 46(10), 4224-4227 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service